2-(Chloromethyl)-5-methyl-3-(methylthio)pyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR300375-250MG
APOSOR300375-250MGEA 1870 EUR
APOSOR300375-250MG
2-(Chloromethyl)-5-methyl-3-(methylthio)pyridine hydrochloride
Formel: C₈H₁₀ClNS·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27924167
CAS-Nummer: 1823869-97-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR