3-Bromo-5-chloro-1H-pyrazolo[3,4-c]pyridine

Lieferant: Apollo Scientific
Achtung

OR300378-1G
APOSOR300378-1GEA 339 EUR
APOSOR300378-1G
3-Bromo-5-chloro-1H-pyrazolo[3,4-c]pyridine
3-Bromo-5-chloro-1H-pyrazolo[3,4-c]pyridine
Formel: C₆H₃BrClN₃
Molecular Weight: 232,465 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13177858
CAS-Nummer: 245325-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR