3-Bromo-4-chloro-1H-pyrazolo[3,4-b]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR300380-1G
APOSOR300380-1GEA
192
EUR
APOSOR300380-1G
3-Bromo-4-chloro-1H-pyrazolo[3,4-b]pyridine
3-Bromo-4-chloro-1H-pyrazolo[3,4-b]pyridine
Formel:
C₆H₃BrClN₃ Molecular Weight: 232,465 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22380452 CAS-Nummer: 1357945-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...