4-Bromoindoline hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR300383-1G
APOSOR300383-1GEA
95.7
EUR
APOSOR300383-1G
4-Bromoindoline hydrochloride
4-Bromoindoline hydrochloride
Formel:
C₈H₈BrN·ClH Molecular Weight: 234,521 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09026827 CAS-Nummer: 1187929-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...