6-Bromo-3-iodo-1H-pyrazolo[4,3-b]pyridine

Lieferant: Apollo Scientific
Achtung

OR300391-250MG
APOSOR300391-250MGEA 98.6 EUR
APOSOR300391-250MG
6-Bromo-3-iodo-1H-pyrazolo[4,3-b]pyridine
6-Bromo-3-iodo-1H-pyrazolo[4,3-b]pyridine
Formel: C₆H₃BrIN₃
Molecular Weight: 323.917 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20923312
CAS-Nummer: 1305208-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR