5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[4,3-b]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR300397-250MG
APOSOR300397-250MGEA
244
EUR
APOSOR300397-250MG
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[4,3-b]pyridine
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[4,3-b]pyridine
Formel:
C₁₁H₁₂BrN₃O Molecular Weight: 282,136 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22689847 CAS-Nummer: 1416712-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...