4-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR300400-1G
OR300400-250MG
APOSOR300400-1GEA
191
EUR
APOSOR300400-1G
APOSOR300400-250MG
4-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine
4-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine
Formel:
C₆H₃BrIN₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD22380433 CAS-Nummer: 1357946-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...