4-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine

Lieferant: Apollo Scientific
Achtung

OR300400-1G OR300400-250MG
APOSOR300400-1GEA 191 EUR
APOSOR300400-1G APOSOR300400-250MG
4-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine
4-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine
Formel: C₆H₃BrIN₃
Lagertemperatur: Kühlschrank
MDL: MFCD22380433
CAS-Nummer: 1357946-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR