2-Hydroxy-4-methylnicotinic acid

Lieferant: Apollo Scientific
Achtung

OR300502-1G OR300502-500MG
APOSOR300502-1GEA 255 EUR
APOSOR300502-1G APOSOR300502-500MG
2-Hydroxy-4-methylnicotinic acid
2-Hydroxy-4-methylnicotinic acid
Formel: C₇H₇NO₃
Schmelzpunkt: 247…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09033815
CAS-Nummer: 38076-81-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR