3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-hydroxy-1H-pyrazole-4-carboxylate

OR300503-1G
APOSOR300503-1GEA 77 EUR
APOSOR300503-1G
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 95%
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester
Formel: C₆H₈N₂O₃
Molecular Weight: 156,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12131138
CAS-Nummer: 7251-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR