Methyl-3-(phenylamino)propanoate

Lieferant: Apollo Scientific

Synonyme: Methyl 3-(phenylamino)propanoate

OR300505-1G OR300505-5G
APOSOR300505-1GEA 132 EUR
APOSOR300505-1G APOSOR300505-5G
Methyl-3-(phenylamino)propanoate
Methyl-3-(phenylamino)propanoate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.2200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00089334
CAS-Nummer: 21911-84-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR