1-Aminocyclohexanecarbonitrile hydrochloride

Lieferant: Apollo Scientific

OR300506-1G OR300506-250MG OR300506-500MG
APOSOR300506-1GEA 0 EUR
APOSOR300506-1G APOSOR300506-250MG APOSOR300506-500MG
1-Aminocyclohexanecarbonitrile hydrochloride
1-Aminocyclohexanecarbonitrile hydrochloride
Formel: C₇H₁₃ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01734623
CAS-Nummer: 50846-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR