1-Aminocyclohexanecarbonitrile hydrochloride
Lieferant: Apollo Scientific
OR300506-1G
OR300506-250MG
OR300506-500MG
APOSOR300506-1GEA
0
EUR
APOSOR300506-1G
APOSOR300506-250MG
APOSOR300506-500MG
1-Aminocyclohexanecarbonitrile hydrochloride
1-Aminocyclohexanecarbonitrile hydrochloride
Formel:
C₇H₁₃ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01734623 CAS-Nummer: 50846-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...