Ethyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat 95%

Lieferant: Apollo Scientific

OR300507-1G
APOSOR300507-1GEA 69.6 EUR
APOSOR300507-1G
Ethyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat 95%
Ethyl-6-bromimidazo[1,2-a]pyridin-3-carboxylat
Formel: C₁₀H₉BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10698068
CAS-Nummer: 372198-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR