4-(Hydroxymethyl)pyrrolidin-2-one
Lieferant: Apollo Scientific
|
Achtung
|
OR300508-1G
APOSOR300508-1GEA
171
EUR
APOSOR300508-1G
4-(Hydroxymethyl)pyrrolidin-2-one
4-(Hydroxymethyl)pyrrolidin-2-one
Formel:
C₅H₉NO₂ Schmelzpunkt: 92…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11845728 CAS-Nummer: 64320-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...