7-Aminopyrazolo[1,5-a]pyrimidine-6-carbonitrile

Lieferant: Apollo Scientific

Synonyme: 7-Amino-6-cyanopyrazolo[1,5-a]pyrimidine

OR300510-1G
APOSOR300510-1GEA 0 EUR
APOSOR300510-1G
7-Aminopyrazolo[1,5-a]pyrimidine-6-carbonitrile
7-Aminopyrazolo[1,5-a]pyrimidine-6-carbonitrile
Formel: C₇H₅N₅
Molecular Weight: 159,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00104599
CAS-Nummer: 89975-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR