6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR300511-1G
APOSOR300511-1GEA 60.9 EUR
APOSOR300511-1G
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid 97%
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₈H₅BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10698063
CAS-Nummer: 944896-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR