3,4-Dihydro-2H-benzo[1,4]oxazine-2-carboxylicacid ethyl ester hydrochloride

Lieferant: Apollo Scientific

Synonyme: Ethyl benzomorpholine-2-carboxylate hydrochloride

OR300517-1G
APOSOR300517-1GEA 0 EUR
APOSOR300517-1G
3,4-Dihydro-2H-benzo[1,4]oxazine-2-carboxylicacid ethyl ester hydrochloride
3,4-Dihydro-2H-benzo[1,4]oxazine-2-carboxylicacid ethyl ester hydrochloride
Formel: C₁₁H₁₄ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11227230
CAS-Nummer: 13582-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR