2-(4-Chlorophenyl)-2-morpholinoacetonitrile
Lieferant: Apollo Scientific
OR300518-1G
APOSOR300518-1GEA
0
EUR
APOSOR300518-1G
2-(4-Chlorophenyl)-2-morpholinoacetonitrile
2-(4-Chlorophenyl)-2-morpholinoacetonitrile
Formel:
C₁₂H₁₃ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173106 CAS-Nummer: 33599-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...