Ethyl-2-chloro-3-oxopropanoate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-chloro-3-oxopropanoate

OR300519-1G
APOSOR300519-1GEA 0 EUR
APOSOR300519-1G
Ethyl-2-chloro-3-oxopropanoate 97%
Ethyl-2-chloro-3-oxopropanoate
Formel: C₅H₇ClO₃
Molecular Weight: 150,56 g/mol
Siedepunkt: 85…90 °C/0,5mmHg
Schmelzpunkt: 20 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07369378
CAS-Nummer: 33142-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR