Methyl-3-(benzylamino)propanoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-(benzylamino)propanoate
OR300523-10G
OR300523-1G
OR300523-5G
OR300523-25G
OR300523-250MG
OR300523-2.5G
APOSOR300523-10GEA
91.5
EUR
APOSOR300523-10G
APOSOR300523-1G
APOSOR300523-5G
APOSOR300523-25G
APOSOR300523-250MG
APOSOR300523-2.5G
Methyl-3-(benzylamino)propanoate
Methyl-3-(benzylamino)propanoate
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193.25 g/mol Siedepunkt: 145…147 °C/7mmHg Lagertemperatur: Gefrierschrank |
MDL:
MFCD01475856 CAS-Nummer: 23574-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...