Ethyl 2-ethyl-5-methylpyrazole-3-carboxylate 95+%

Lieferant: Apollo Scientific

OR300543-1G
APOSOR300543-1GEA 139 EUR
APOSOR300543-1G
Ethyl 2-ethyl-5-methylpyrazole-3-carboxylate 95+%
Ethyl 2-ethyl-5-methylpyrazole-3-carboxylate
Formel: C₉H₁₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00159650
CAS-Nummer: 50920-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR