Methyl 3-[N-tert-butoxycarbonylmethoxycarbonyl)methylamino]propanoate

Lieferant: Apollo Scientific

Synonyme: 3-(tert-Butoxycarbonylmethoxycarbonylmethylamino)propanoic acid methyl ester

OR300551-1G OR300551-100MG OR300551-250MG OR300551-500MG OR300551-2.5G
APOSOR300551-1GEA 275 EUR
APOSOR300551-1G APOSOR300551-100MG APOSOR300551-250MG APOSOR300551-500MG APOSOR300551-2.5G
Methyl 3-[N-tert-butoxycarbonylmethoxycarbonyl)methylamino]propanoate
Methyl 3-[N-tert-butoxycarbonylmethoxycarbonyl)methylamino]propanoate
Formel: C₁₂H₂₁NO₆
Molecular Weight: 275.298 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369713
CAS-Nummer: 194924-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR