5-Bromo-4,6-dimethyl-2-hydroxynicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-4,6-dimethyl-2-hydroxypyridine-3-carboxylic acid, 3-Bromo-5-carboxy-2,4-dimethyl-6-hydroxypyridine
OR300592-1G
APOSOR300592-1GEA
0
EUR
APOSOR300592-1G
5-Bromo-4,6-dimethyl-2-hydroxynicotinic acid
5-Bromo-4,6-dimethyl-2-hydroxynicotinic acid
Formel:
C₈H₈BrNO₃ Molecular Weight: 246,058 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19981490 CAS-Nummer: 339366-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...