3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl N-Boc-3-oxopyrrolidine-2-carboxylate, 1-tert-Butyl 2-ethyl 3-oxopyrrolidine-1,2-dicarboxylate

OR300597-1G
APOSOR300597-1GEA 192 EUR
APOSOR300597-1G
3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester
3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester
Formel: C₁₂H₁₉NO₅
Molecular Weight: 257.29 g/mol
Siedepunkt: 150 °C/0.375mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD11052440
CAS-Nummer: 170123-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR