5-Methyl-2-[4-(nitrophenyl)-1,3,4-oxadiazole

Lieferant: Apollo Scientific
Achtung

OR300601-1G
APOSOR300601-1GEA 348 EUR
APOSOR300601-1G
5-Methyl-2-[4-(nitrophenyl)-1,3,4-oxadiazole
5-Methyl-2-[4-(nitrophenyl)-1,3,4-oxadiazole
Formel: C₉H₇N₃O₃
Molecular Weight: 205,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00186437
CAS-Nummer: 22815-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR