5-Oxopyrrolidine-3-carboxylic acid amide

Lieferant: Apollo Scientific

Synonyme: 5-Oxopyrrolidine-3-carboxamide, Pyrrolidin-5-one-3-carboxamide

OR300643-1G
APOSOR300643-1GEA 42.2 EUR
APOSOR300643-1G
5-Oxopyrrolidine-3-carboxylic acid amide
5-Oxopyrrolidine-3-carboxylic acid amide
Formel: C₅H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03613706
CAS-Nummer: 14466-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR