Morpholin-4-yl(phenyl)acetonitrile

Lieferant: Apollo Scientific

OR300647-1G OR300647-250MG OR300647-5G OR300647-10G OR300647-25G
APOSOR300647-1GEA 0 EUR
APOSOR300647-1G APOSOR300647-250MG APOSOR300647-5G APOSOR300647-10G APOSOR300647-25G
Morpholin-4-yl(phenyl)acetonitrile
Morpholin-4-yl(phenyl)acetonitrile
Formel: C₁₂H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00014619
CAS-Nummer: 15190-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR