tert-Butyl-2-formylpiperidine-1-carboxylate 97%

Lieferant: Apollo Scientific

OR300652-25G
APOSOR300652-25GEA 127 EUR
APOSOR300652-25G
tert-Butyl-2-formylpiperidine-1-carboxylate 97%
tert-Butyl-2-formylpiperidine-1-carboxylate
Formel: C₁₁H₁₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02179021
CAS-Nummer: 157634-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR