2-Chloro-4-phenylnicotinonitrile 97%

Lieferant: Apollo Scientific

OR300654-5G
APOSOR300654-5GEA 0 EUR
APOSOR300654-5G
2-Chloro-4-phenylnicotinonitrile 97%
2-Chloro-4-phenylnicotinonitrile
Formel: C₁₂H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01935958
CAS-Nummer: 163563-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR