6-Methoxy-1H-indole-2-carboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 6-Methoxyindole-2-carboxylic acid

OR300659-5G
APOSOR300659-5GEA 0 EUR
APOSOR300659-5G
6-Methoxy-1H-indole-2-carboxylic acid 98%
6-Methoxy-1H-indole-2-carboxylic acid
Formel: C₁₀H₉NO₃
Molecular Weight: 191,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01548823
CAS-Nummer: 16732-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR