Boc-Tic-OH 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-Tic-OH, (S)-2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

OR30067-5G
APOSOR30067-5GEA 21.9 EUR
APOSOR30067-5G
Boc-Tic-OH 98%
Boc-Tic-OH
Formel: C₁₅H₁₉NO₄
Molecular Weight: 277.32 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00143845
CAS-Nummer: 78879-20-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR