2-(Methylthio)pyrimidine-4,6-diol 98+%

Lieferant: Apollo Scientific

Synonyme: 2-Methylsulphanyl-pyrimidine-4,6-diol, 4,6-Dihydroxy-2-methylsulphanylpyrimidine

OR300689-100G
APOSOR300689-100GEA 62.5 EUR
APOSOR300689-100G
2-(Methylthio)pyrimidine-4,6-diol 98+%
2-(Methylthio)pyrimidine-4,6-diol
Formel: C₅H₆N₂O₂S
Molecular Weight: 158.18 g/mol
MDL: MFCD00006087
CAS-Nummer: 29639-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR