2-Phenylthiazole-5-carboxylic acid 97%

Lieferant: Apollo Scientific

OR300695-250MG
APOSOR300695-250MGEA 104 EUR
APOSOR300695-250MG
2-Phenylthiazole-5-carboxylic acid 97%
2-Phenylthiazole-5-carboxylic acid
Formel: C₁₀H₇NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD07376773
CAS-Nummer: 10058-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR