3-Hydroxy-5-phenylcyclohex-2-enone 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR300698-10G
APOSOR300698-10GEA
282
EUR
APOSOR300698-10G
3-Hydroxy-5-phenylcyclohex-2-enone 95+%
3-Hydroxy-5-phenylcyclohex-2-enone
Formel:
C₁₂H₁₂O₂ Molecular Weight: 188,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463813 CAS-Nummer: 35376-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...