4-Amino-1-isoindolinone

Lieferant: Apollo Scientific

Synonyme: 4-Amino-2,3-dihydro-1H-isoindol-1-one

OR300702-1G
APOSOR300702-1GEA 85.7 EUR
APOSOR300702-1G
4-Amino-1-isoindolinone
4-Amino-1-isoindolinone
Formel: C₈H₈N₂O
Molecular Weight: 148,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234883
CAS-Nummer: 366452-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR