Ethyl-5-methyl[1,2,4]oxadiazole-3-carboxylate
Lieferant: Apollo Scientific
OR300717-1G
APOSOR300717-1GEA
89.9
EUR
APOSOR300717-1G
Ethyl-5-methyl[1,2,4]oxadiazole-3-carboxylate
Ethyl-5-methyl[1,2,4]oxadiazole-3-carboxylate
Formel:
C₆H₈N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864941 CAS-Nummer: 40699-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...