5-Amino-3-methyl-isothiazole-4-carbonitrile

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Amino-4-cyano-3-methylisothiazole, 5-Amino-3-methylisothiazole-4-carbonitrile

OR300718-1G
APOSOR300718-1GEA 0 EUR
APOSOR300718-1G
5-Amino-3-methyl-isothiazole-4-carbonitrile
5-Amino-3-methyl-isothiazole-4-carbonitrile
Formel: C₅H₅N₃S
Molecular Weight: 139,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422694
CAS-Nummer: 41808-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR