Ethyl-2,3-dihydro-1,4-benzodioxin-2-carboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 1,4-Benzodioxane-2-carboxylate, Ethyl 2,3-dihydrobenzo[1,4]dioxine-2-carboxylate, 1,4-Benzodioxane-2-carboxylic acid ethyl ester
OR300724-5G
APOSOR300724-5GEA
43.5
EUR
APOSOR300724-5G
Ethyl-2,3-dihydro-1,4-benzodioxin-2-carboxylat 95%
Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate
Formel:
C₁₁H₁₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01099774 CAS-Nummer: 4739-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...