1,3-Dihydroindole-2-thione

Lieferant: Apollo Scientific
Achtung

OR300726-1G
APOSOR300726-1GEA 0 EUR
APOSOR300726-1G
1,3-Dihydroindole-2-thione
1,3-Dihydroindole-2-thione
Formel: C₈H₇NS
Lagertemperatur: Kühlschrank
MDL: MFCD00129941
CAS-Nummer: 496-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR