2-Amino-5-iodonicotinic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-iodopyridine-3-carboxylic acid

OR300739-5G
APOSOR300739-5GEA 243 EUR
APOSOR300739-5G
2-Amino-5-iodonicotinic acid 95+%
2-Amino-5-iodonicotinic acid
Formel: C₆H₅IN₂O₂
Molecular Weight: 264,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568356
CAS-Nummer: 54400-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR