Ethylpyrazol-3-carboxylat 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: Pyrazole-3-carboxylic Acid Ethyl Ester, Ethyl 1H-pyrazole-3-carboxylate

OR300747-25G
APOSOR300747-25GEA 37.7 EUR
APOSOR300747-25G
Ethylpyrazol-3-carboxylat 99%
Ethyl pyrazole-3-carboxylate
Formel: C₆H₈N₂O₂
Schmelzpunkt: 159…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00159643
CAS-Nummer: 5932-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR