1-Cyano-1-(phenylamino)cyclohexane

Lieferant: Apollo Scientific

Synonyme: 1-Phenylaminocyclohexanecarbonitrile

OR300759-1G
APOSOR300759-1GEA 166 EUR
APOSOR300759-1G
1-Cyano-1-(phenylamino)cyclohexane
1-Cyano-1-(phenylamino)cyclohexane
Formel: C₁₃H₁₆N₂
Molecular Weight: 200,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08166730
CAS-Nummer: 64269-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR