3-Pyridyloxyessigsäure 98+%
Lieferant: Apollo Scientific
|
Achtung
|
OR300774-1G
APOSOR300774-1GEA
145
EUR
APOSOR300774-1G
3-Pyridyloxyessigsäure 98+%
3-Pyridyloxyessigsäure
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039772 CAS-Nummer: 86649-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...