2-Phenyl-2-(piperidin-1-yl)acetonitrile 97%

Lieferant: Apollo Scientific

Synonyme: α-(Piperidin-1-yl)phenylacetonitrile

OR300829-1G
APOSOR300829-1GEA 132 EUR
APOSOR300829-1G
2-Phenyl-2-(piperidin-1-yl)acetonitrile 97%
2-Phenyl-2-(piperidin-1-yl)acetonitrile
Formel: C₁₃H₁₆N₂
Molecular Weight: 200,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023711
CAS-Nummer: 5766-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR