4-(2-Hydroxyethyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR30086-1G OR30086-250MG
APOSOR30086-1GEA 34.8 EUR
APOSOR30086-1G APOSOR30086-250MG
4-(2-Hydroxyethyl)benzonitril 98%
4-(2-Hydroxyethyl)benzonitril
Formel: C₉H₉NO
Molecular Weight: 147.18 g/mol
Siedepunkt: 116…118 °C/0.5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00052461
CAS-Nummer: 69395-13-7
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR