Methyl-2-morpholino-2-phenylacetate hydrochloride

Lieferant: Apollo Scientific

OR300892-1G
APOSOR300892-1GEA 0 EUR
APOSOR300892-1G
Methyl-2-morpholino-2-phenylacetate hydrochloride
Methyl-2-morpholino-2-phenylacetate hydrochloride
Formel: C₁₃H₁₈ClNO₃
Molecular Weight: 271,75 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28099762
CAS-Nummer: 27594-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR