3-Methyl-2-(morpholin-4-yl)butanenitrile hydrochloride

Lieferant: Apollo Scientific

OR300896-1G
APOSOR300896-1GEA 0 EUR
APOSOR300896-1G
3-Methyl-2-(morpholin-4-yl)butanenitrile hydrochloride
3-Methyl-2-(morpholin-4-yl)butanenitrile hydrochloride
Formel: C₉H₁₇ClN₂O
Molecular Weight: 204,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28099771
CAS-Nummer: 1672675-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR