Ethyl 2-(4-bromophenyl)thio-2-methylpropionate

Lieferant: Apollo Scientific

OR300897-1G
APOSOR300897-1GEA 0 EUR
APOSOR300897-1G
Ethyl 2-(4-bromophenyl)thio-2-methylpropionate
Ethyl 2-(4-bromophenyl)thio-2-methylpropionate
Formel: C₁₂H₁₅BrO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD28122845
CAS-Nummer: 18527-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR