4-Benzylpiperidine-1-carboxamidine acetate
Lieferant: Apollo Scientific
OR300898-1G
APOSOR300898-1GEA
0
EUR
APOSOR300898-1G
4-Benzylpiperidine-1-carboxamidine acetate
4-Benzylpiperidine-1-carboxamidine acetate
Formel:
C₁₃H₁₉N₃·CH₃CO₂H Molecular Weight: 277,362 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28122846 CAS-Nummer: 1672675-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...